Assay Details
Gene Symbol: CRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493542.6 | XP_001493592.3 | 423 | 63 | ||
XM_023655538.1 | XP_023511306.1 | 459 | 63 | |||
XM_023655539.1 | XP_023511307.1 | 348 | 63 | |||
XM_023655540.1 | XP_023511308.1 | 401 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
