Assay Details
Gene Symbol: BCL7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598940.3 | XP_005598997.2 | 697 | 91 | ||
XM_023655612.1 | XP_023511380.1 | 697 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
