Assay Details
Gene Symbol: IL21R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598826.3 | XP_005598883.1 | 1419 | 71 | ||
XM_005598827.2 | XP_005598884.1 | 866 | 71 | |||
XM_005598828.3 | XP_005598885.1 | 1803 | 71 | |||
XM_023616084.1 | XP_023471852.1 | 1725 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
