Assay Details
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598856.3 | XP_005598913.1 | 502 | 92 | ||
XM_023616136.1 | XP_023471904.1 | 750 | 92 | |||
XM_023616137.1 | XP_023471905.1 | 750 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
