Assay Details
Gene Symbol: GDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488082.4 | XP_001488132.1 | 984 | 70 | ||
XM_005598882.3 | XP_005598939.1 | 951 | 70 | |||
XM_023616229.1 | XP_023471997.1 | 806 | 70 | |||
GenBank mRNA | JL626958.1 | - | 4 - 5 | 572 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
