Assay Details
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488163.5 | XP_001488213.3 | 658 | 69 | ||
XM_023616255.1 | XP_023472023.1 | 585 | 69 | |||
XM_023616256.1 | XP_023472024.1 | 1044 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
