Assay Details
Gene Symbol: ARL6IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488398.5 | XP_001488448.2 | 612 | 63 | ||
GenBank mRNA | JL616132.2 | - | 5 - 6 | 498 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
