Assay Details
Gene Symbol: MPV17L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616280.1 | XP_023472048.1 | 446 | 60 | ||
XM_023616282.1 | XP_023472050.1 | 373 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
