Assay Details
Gene Symbol: MRTFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598901.3 | XP_005598958.1 | 5316 | 68 | ||
XM_023616297.1 | XP_023472065.1 | 1853 | 68 | |||
XM_023616298.1 | XP_023472066.1 | 1866 | 68 | |||
XM_023616299.1 | XP_023472067.1 | 555 | 68 | |||
XM_023616300.1 | XP_023472068.1 | 492 | 68 | |||
XM_023616301.1 | XP_023472069.1 | 1867 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
