Assay Details
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599099.3 | XP_005599156.2 | 1470 | 57 | ||
XM_023616357.1 | XP_023472125.1 | 948 | 57 | |||
XM_023616358.1 | XP_023472126.1 | 1210 | 57 | |||
XM_023616359.1 | XP_023472127.1 | 867 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
