Assay Details
Gene Symbol: KCTD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014730138.2 | XP_014585624.2 | 499 | 62 | ||
XM_023616534.1 | XP_023472302.1 | 499 | 62 | |||
XM_023616535.1 | XP_023472303.1 | 497 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
