Assay Details
Gene Symbol: ECI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616570.1 | XP_023472338.1 | 666 | 58 | ||
XM_023616571.1 | XP_023472339.1 | 502 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
