Assay Details
Gene Symbol: NTHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915338.4 | XP_001915373.2 | 725 | 97 | ||
XM_023616593.1 | XP_023472361.1 | 800 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
