Assay Details
Gene Symbol: SLC9A3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616587.1 | XP_023472355.1 | 572 | 93 | ||
XM_023616588.1 | XP_023472356.1 | 572 | 93 | |||
XM_023616589.1 | XP_023472357.1 | 572 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
