Assay Details
Gene Symbol: ZNF598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616594.1 | XP_023472362.1 | 1251 | 55 | ||
XM_023616595.1 | XP_023472363.1 | 1242 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
