Assay Details
Gene Symbol: NOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616597.1 | XP_023472365.1 | 811 | 58 | ||
XM_023616598.1 | XP_023472366.1 | 1004 | 58 | |||
XM_023616599.1 | XP_023472367.1 | 1122 | 58 | |||
XM_023616600.1 | XP_023472368.1 | 1031 | 58 | |||
XM_023616601.1 | XP_023472369.1 | 907 | 58 | |||
XM_023616603.1 | XP_023472371.1 | 763 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
