Assay Details
Gene Symbol: NUBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616640.1 | XP_023472408.1 | 794 | 64 | ||
XM_023616641.1 | XP_023472409.1 | 733 | 64 | |||
XM_023616642.1 | XP_023472410.1 | 1532 | 64 | |||
XM_023616643.1 | XP_023472411.1 | 820 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
