Assay Details
Gene Symbol: UNKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014730265.2 | XP_014585751.1 | 795 | 52 | ||
XM_023616664.1 | XP_023472432.1 | 1915 | 52 | |||
XM_023616666.1 | XP_023472434.1 | 2099 | 52 | |||
XM_023616667.1 | XP_023472435.1 | 2187 | 52 | |||
XM_023616668.1 | XP_023472436.1 | 1969 | 52 | |||
XM_023616669.1 | XP_023472437.1 | 794 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
