Assay Details
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495724.4 | XP_001495774.2 | 1174 | 96 | ||
XM_005599085.3 | XP_005599142.1 | 1179 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
