Assay Details
Gene Symbol: NME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616743.1 | XP_023472511.1 | 372 | 79 | ||
XM_023616744.1 | XP_023472512.1 | 372 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
