Assay Details
Gene Symbol: FAM234A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496735.3 | XP_001496785.1 | 1611 | 58 | ||
XM_005599092.3 | XP_005599149.1 | 1611 | 58 | |||
XM_014730297.2 | XP_014585783.1 | 1579 | 58 | |||
XM_014730298.2 | XP_014585784.1 | 1456 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
