Assay Details
Gene Symbol: EFCAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023616980.1 | XP_023472748.1 | 398 | 80 | ||
XM_023616982.1 | XP_023472750.1 | 539 | 80 | |||
XM_023616984.1 | XP_023472752.1 | 463 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
