Assay Details
Gene Symbol: CREBRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502910.6 | XP_001502960.3 | 443 | 86 | ||
XM_023617224.1 | XP_023472992.1 | 716 | 86 | |||
XM_023617225.1 | XP_023472993.1 | 598 | 86 | |||
XM_023617226.1 | XP_023472994.1 | 611 | 86 | |||
XM_023617227.1 | XP_023472995.1 | 410 | 86 | |||
XM_023617228.1 | XP_023472996.1 | 457 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
