Assay Details
Gene Symbol: GABRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500005.6 | XP_001500055.3 | 777 | 68 | ||
XM_014730527.2 | XP_014586013.2 | 863 | 68 | |||
XM_023617259.1 | XP_023473027.1 | 915 | 68 | |||
XM_023617260.1 | XP_023473028.1 | 467 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
