Assay Details
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503154.5 | XP_001503204.1 | 624 | 76 | ||
XM_014730528.2 | XP_014586014.1 | 655 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
