Assay Details
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599241.3 | XP_005599298.1 | 135 | 90 | ||
XM_005599243.3 | XP_005599300.1 | 136 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005599241.3, XM_005599243.3 transcripts.
Set Membership: |
![]() ![]() |
