Assay Details
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503444.5 | XP_001503494.1 | 445 | 117 | ||
XM_005599239.3 | XP_005599296.1 | 407 | 117 | |||
XM_005599240.3 | XP_005599297.1 | 380 | 117 | |||
XM_005599241.3 | XP_005599298.1 | 436 | 117 | |||
XM_005599243.3 | XP_005599300.1 | 434 | 117 | |||
XM_023617294.1 | XP_023473062.1 | 379 | 117 |

More Information
Set Membership: |
![]() ![]() |
