Assay Details
Gene Symbol: TIMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023617338.1 | XP_023473106.1 | 765 | 62 | ||
XM_023617339.1 | XP_023473107.1 | 762 | 62 | |||
XM_023617340.1 | XP_023473108.1 | 765 | 62 | |||
XM_023617341.1 | XP_023473109.1 | 765 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
