Assay Details
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599291.3 | XP_005599348.1 | 460 | 66 | ||
XM_014730574.2 | XP_014586060.1 | 677 | 66 | |||
XM_014730575.2 | XP_014586061.1 | 672 | 66 | |||
XM_023617406.1 | XP_023473174.1 | 813 | 66 | |||
XM_023617407.1 | XP_023473175.1 | 469 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
