Assay Details
Gene Symbol: IL17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503774.4 | XP_001503824.1 | 141 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001503774.4 transcripts.
Set Membership: |
![]() ![]() |
