Assay Details
Gene Symbol: IL17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503774.4 | XP_001503824.1 | 434 | 114 | ||
XM_023617455.1 | XP_023473223.1 | 436 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
