Assay Details
Gene Symbol: PCYOX1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503777.4 | XP_001503827.2 | 749 | 53 | ||
XM_023617460.1 | XP_023473228.1 | 774 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
