Assay Details
Gene Symbol: CDKN2AIPNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599422.3 | XP_005599479.1 | 311 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
