Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023617751.1 | XP_023473519.1 | 1018 | 110 | ||
XM_023617752.1 | XP_023473520.1 | 706 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
