Assay Details
Gene Symbol: FEZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739216.2 | XP_014594702.1 | 1913 | 99 | ||
XM_023639565.1 | XP_023495333.1 | 1816 | 99 | |||
XM_023639566.1 | XP_023495334.1 | 1674 | 99 | |||
XM_023639567.1 | XP_023495335.1 | 1665 | 99 | |||
XM_023639568.1 | XP_023495336.1 | 1901 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
