Assay Details
Gene Symbol: WDR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645320.1 | XP_023501088.1 | 631 | 115 | ||
XM_023645321.1 | XP_023501089.1 | 547 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
