Assay Details
Gene Symbol: LOC100061656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740695.2 | XP_014596181.1 | 3417 | 85 | ||
XM_014740696.2 | XP_014596182.1 | 3417 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
