Assay Details
Gene Symbol: NCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642552.1 | XP_023498320.1 | 2333 | 97 | ||
GenBank mRNA | AY246725.1 | - | 2 | 116 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
