Assay Details
Gene Symbol: TNFAIP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503409.6 | XP_001503459.2 | 1656 | 118 | ||
XM_005599492.3 | XP_005599549.1 | 1516 | 118 | |||
XM_005599493.3 | XP_005599550.1 | 1480 | 118 | |||
XM_005599495.3 | XP_005599552.1 | 1818 | 118 | |||
XM_014730723.2 | XP_014586209.1 | 1518 | 118 | |||
XM_023617828.1 | XP_023473596.1 | 1748 | 118 | |||
GenBank mRNA | JL616267.1 | - | 1 | 1367 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
