Assay Details
Gene Symbol: LOC111775214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649877.1 | XP_023505645.1 | 715 | 83 | ||
XM_023649878.1 | XP_023505646.1 | 959 | 83 | |||
XM_023649879.1 | XP_023505647.1 | 921 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
