Assay Details
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364972.4 | XP_003365020.2 | 646 | 80 | ||
XM_014739636.2 | XP_014595122.2 | 646 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
