Assay Details
Gene Symbol: NXPE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003365321.2 | XP_003365369.1 | 930 | 85 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NXPE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003365321.2 | XP_003365369.1 | 930 | 85 |