Assay Details
Gene Symbol: EIF4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597755.3 | XP_005597812.1 | 432 | 73 | ||
XM_023653459.1 | XP_023509227.1 | 366 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
