Assay Details
Gene Symbol: TRAPPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597759.2 | XP_005597816.1 | 291 | 72 | ||
XM_023653744.1 | XP_023509512.1 | 283 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
