Assay Details
Gene Symbol: PRSS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598747.3 | XP_005598804.1 | 2319 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
