Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599055.3 | XP_005599112.1 | 1005 | 57 | ||
XM_005599056.3 | XP_005599113.1 | 951 | 57 | |||
XM_005599057.3 | XP_005599114.1 | 1005 | 57 | |||
XM_014730233.2 | XP_014585719.1 | 1005 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
