Assay Details
Gene Symbol: UBAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602994.3 | XP_005603051.2 | 3582 | 90 | ||
XM_023655359.1 | XP_023511127.1 | 2030 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
