Assay Details
Gene Symbol: RBM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603222.3 | XP_005603279.1 | 792 | 74 | ||
XM_014733501.2 | XP_014588987.1 | 792 | 74 | |||
XM_014733502.2 | XP_014588988.1 | 889 | 74 | |||
XM_014733503.2 | XP_014588989.1 | 1010 | 74 | |||
XM_014733504.2 | XP_014588990.1 | 689 | 74 | |||
XM_014733505.2 | XP_014588991.1 | 795 | 74 | |||
XM_023622050.1 | XP_023477818.1 | 1253 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
