Assay Details
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606682.3 | XP_005606739.1 | 532 | 117 | ||
XM_005606683.3 | XP_005606740.1 | 543 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
