Assay Details
Gene Symbol: LOC100060483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607698.3 | XP_005607755.1 | 278 | 93 | ||
XM_005607699.3 | XP_005607756.1 | 278 | 93 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100060483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607698.3 | XP_005607755.1 | 278 | 93 | ||
XM_005607699.3 | XP_005607756.1 | 278 | 93 |